Order Entry
Austria
ContactUsLinkComponent
Lugol's iodine solution, Sigma-Aldrich®
Lugol's iodine solution, Sigma-Aldrich®
Certificates
Lugol's iodine solution, Sigma-Aldrich®
Certificates

Specification Test Results

  • Appearance (colour):
    Report result
  • Appearance (form):
    Report result
  • Argentometr. titration:
    Assay potassium iodide (KI)
  • Redox titration:
    Assay iodine (i)
  • Redox titration (method):
    Titration with sodium thiosulfate
  • Density d20/4:
    Report result
  • Recommended retest period:
    42 months

Specification Test Results

Related Information

Frequently Bought Together