Order Entry
Austria
ContactUsLinkComponent
Immersion oil for microscopy, Sigma-Aldrich®
Immersion oil for microscopy, Sigma-Aldrich®
Supplier:  Merck
Certificates
Immersion oil for microscopy, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specifications

  • M-Clarity quality level:
    MQ400
  • Viscosity:
    100 - 120 mPa∙s (20 °C)
  • Density:
    1.02 g/cm3 (20 °C)

Specifications

Frequently Bought Together