Order Entry
Austria
ContactUsLinkComponent
Haemalum (Mayer's), Sigma-Aldrich®
Health
Irritant
Haemalum (Mayer's), Sigma-Aldrich®
Supplier:  Merck
Certificates
undefined
Health
Irritant
Haemalum (Mayer's), Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Density:
    1.05 g/cm3 (20 °C)

Specification Test Results

Frequently Bought Together