May Grunwald's eosin-methylene blue modified solution stain for hematology, for cytology, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
:
MDL Number:MFCD00131580
Density:0.79 g/cm³ (20 °C)
ADR:3 (6.1),II
:M-Clarity™ quality level = MQ300
UN:1230
:GHS02
:GHS06
:GHS08
:1
Limited Time Sale
Select items below are currently on sale
Specification Test Results
- Suitability for microscopy (Blood smear):passes test
- Erythrocytes:pink to brownish
- Nuclei:violet
- Eosinophilic granules:red-brown
- Neutrophilic granules:light violet
- Lymphocyte cytoplasm:Blue
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.01424.0100_TDS_EN MERC1.01424.0500_TDS_EN MERC1.01424.1000_TDS_EN MERC1.01424.2500_TDS_EN ProductPage_101424
Product specification
Merck The Color of Tissue Diagnostics
Product Literature
Frequently Bought Together