


May Grunwald's eosin-methylene blue solution, Sigma-Aldrich®
Catalog # MG1L-1L
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
ADR:3,II
:M-Clarity™ quality level = MQ500 (more info.)
UN:1230
:GHS02
:GHS06
:GHS08
Storage Temperature:Ambient
:1
MDL Number:MFCD00131580