Order Entry
Austria
ContactUsLinkComponent
May Grunwald's eosin-methylene blue in methanol, Sigma-Aldrich®
Irritant
May Grunwald's eosin-methylene blue in methanol, Sigma-Aldrich®
Catalog # 32855-25G
CAS Number:  
Irritant
May Grunwald's eosin-methylene blue in methanol, Sigma-Aldrich®
Catalog # 32855-25G
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Solubility:
    0.01 g in 10 ml methanol
  • Wavelength (1) (UV):
    635.0 - 655.0 nm
  • Wavelength (2) (UV):
    512.0 - 532.0 nm
  • Extinction (1%/1 cm) (1) (UV):
    ≥1000
  • Extinction (1%/1 cm) (2) (UV):
    ≥650

Specification Test Results

About this item

 :
 :M-Clarity™ quality level = MQ200
 :GHS07
Storage Temperature:Ambient
MDL Number:MFCD00131580