Order Entry
Austria
ContactUsLinkComponent
May Grunwald's eosin-methylene blue
Irritant
May Grunwald's eosin-methylene blue
Catalog # MOLEM35755821
Supplier:  Molekula
CAS Number:  
Irritant
May Grunwald's eosin-methylene blue
Catalog # MOLEM35755821
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :GHS07
Storage Temperature:Ambient
MDL Number:MFCD00131580