Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
May Grunwald's eosin-methylene blue
May Grunwald's eosin-methylene blue
May Grunwald's eosin-methylene blue
Irritant
May Grunwald's eosin-methylene blue
Catalog # CTEH61024/1
Supplier:
CLIN TECH
CAS Number:
Expand image
Irritant
May Grunwald's eosin-methylene blue
Catalog # CTEH61024/1
Supplier:
CLIN TECH
CAS Number:
Specifications
Pack type:
Glass bottle
Pk:
1 L
Seq:
5037
Specifications
About this item
Product Details
:
:
GHS07
Storage Temperature:
Ambient
MDL Number:
MFCD00131580
Product Details