
Giemsa's stain for microscopy, Sigma-Aldrich®
Lieferant: Merck
Zertifikate
Über diesen Artikel
CAS-Nummer:51811-82-6
:
MDL-Nummer:MFCD00081642
:M-Clarity™ quality level = MQ400
:GHS07
:1
Spezifikation Testergebnisse
- Identity (UV/VIS-Spectrum):passes test
- Loss on drying (105 °C):≤ 10 %
- Suitability for microscopy (Blood smear):passes test
- Nuclei:violet
- Lymphocytes (Plasma):Blue
- Monocytes (Plasma):grey blue
- basophilic granulocytes (Granules):dark violet
- eosinophilic granulocytes (Granules):reddish to red-brown
- Neutrophilic granulocytes (Granules):red-violet
- Thrombocytes:violet
- Erythrocytes:reddish-brownish
- Blood parasites (Nuclei):lightred
Spezifikation Testergebnisse
Zugehörige Informationen
Zertifikate
Suche nach ChargennummerMerck The Color of Tissue Diagnostics
Product Literature
msds_109203
SDS
ProductPage_109203 ProductPage_109203
Product specification
Frequently Bought Together