
Giemsa's stain, certified by the Biological Stain Commission, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:51811-82-6
:
MDL Number:MFCD00081642
Melting Pt:300 °C (lit.)
:M-Clarity™ quality level = MQ200
EINECS:257-438-2
:GHS07
Storage Temperature:Room Temperature
Frequently Bought Together