Eukitt® mounting medium
About this item
This is a mounting medium (contains 45% acrylic resin and 55% xylenes) used in microscopy. It is an adhesive and specimen preservative that can be used manually and in automated coverslipping equipment. Special purpose adhesive and sealant for use with glass, non plastic and other non-reactive materials.
- Fast-drying (20 minutes)
- Refractive index close to glass (1.510)
- Chemically neutral, little sensitivity to water, no reaction to staining
- Temperature limits: –17 to 42 °C
- Not affected by ultraviolet light, non yellowing, stays optically clear for over 10 years
- Spreads quickly and evenly without forming any air bubbles
Can be used in place of epoxy resins for mounting petrographic thin sections, used in fluorescence dyes in bones.
2 Options Available Below
Product Details & Documents
This is a mounting medium (contains 45% acrylic resin and 55% xylenes) used in microscopy. It is an adhesive and specimen preservative that can be used manually and in automated coverslipping equipment. Special purpose adhesive and sealant for use with glass, non plastic and other non-reactive materials.
- Fast-drying (20 minutes)
- Refractive index close to glass (1.510)
- Chemically neutral, little sensitivity to water, no reaction to staining
- Temperature limits: –17 to 42 °C
- Not affected by ultraviolet light, non yellowing, stays optically clear for over 10 years
- Spreads quickly and evenly without forming any air bubbles
Can be used in place of epoxy resins for mounting petrographic thin sections, used in fluorescence dyes in bones.
Certificates
Search by Lot NumberSpecifications for Product Family
Specifications
Specifications
- Catalog No:
- 03989-100ML
- 03989-500ML
- M-Clarity quality level:
- MQ100
- MQ100
Specifications
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- 03989-100MLEukitt® mounting mediumSpecifications:
More
Specifications
Cat. No.03989-100MLM-Clarity quality levelMQ100 - 03989-500MLEukitt® mounting mediumSpecifications:
More
Specifications
Cat. No.03989-500MLM-Clarity quality levelMQ100
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen.