Orange G, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:1936-15-8
:
Formula:C₁₆H₁₀N₂Na₂O₇S₂
MDL Number:MFCD00012457
:452.38 g/mol
Melting Pt:141 °C
Density:0.45…0.47 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:217-705-6
Frequently Bought Together