Order Entry
Austria
ContactUsLinkComponent
Bismarck brown R for microscopy, Sigma-Aldrich®
Bismarck brown R for microscopy, Sigma-Aldrich®
Catalog # 15000-100G
CAS Number:  
Bismarck brown R for microscopy, Sigma-Aldrich®
Catalog # 15000-100G
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Basic Brown 4, Vesuvine, 1,3-Di((2,4-diamino-5-methylphenyl)azo)-4-methylbenzol, C.I. 21010
 :
 :461.39 g/mol
EINECS:226-541-4
CAS Number:5421-66-9
 :1052-38-6
Formula:C₂₁H₂₄N₈·2HCl
MDL Number:MFCD00036386