2,6-Dichlorophenolindophenol sodium salt dihydrate ≥98% (dry basis), GR for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:620-45-1
:1266615-56-8
Formula:C₁₂H₆Cl₂NNaO₂·2H₂O
MDL Number:MFCD00150015
:326.105 g/mol
Melting Pt:? 300 °C
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:210-640-4
Specification Test Results
- Solubility:(20 °C) slightly soluble
- Molar Mass:326.11 g/mol
- Bulk Density:320 kg/m3
Specification Test Results
Frequently Bought Together