2,6-Dichlorophenolindophenol sodium salt hydrate
Catalog # MOLE10933059
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:290.08 g/mol (anhy)
Melting Pt:> 300 °C
EINECS:210-640-4
CAS Number:1266615-56-8
:620-45-1
Formula:C₁₂H₆Cl₂NNaO₂·xH₂O
MDL Number:MFCD00150014