2,6-Dichlorophenolindophenol sodium salt hydrate, BioReagent for the determination of vitamin C, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:266615-56-8
:620-45-1
Formula:C₁₂H₆Cl₂NNaO₂·xH₂O
MDL Number:MFCD00150014
:290.08 g/mol (anhy)
Melting Pt:> 300 °C
EINECS:210-640-4
Frequently Bought Together