Order Entry
Austria
ContactUsLinkComponent
Schiff reagent (Feulgen stain) for analysis of aldehydes
Schiff reagent (Feulgen stain) for analysis of aldehydes
Catalog # CTEH610280
Supplier:  CLIN TECH
CAS Number:  
Schiff reagent (Feulgen stain) for analysis of aldehydes
Catalog # CTEH610280
Supplier:  CLIN TECH
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
Density:1.1 g/cm³ (20 °C)
MDL Number:MFCD00081985