Order Entry
Austria
ContactUsLinkComponent
Carbol fuchsin (Ziehl-Neelsen) for microscopy and bacteriology phenol-free, Sigma-Aldrich®
Flammable
Corrosive
Health
Irritant
Carbol fuchsin (Ziehl-Neelsen) for microscopy and bacteriology phenol-free, Sigma-Aldrich®
Certificates
Flammable
Corrosive
Health
Irritant
Carbol fuchsin (Ziehl-Neelsen) for microscopy and bacteriology phenol-free, Sigma-Aldrich®
Certificates

Specification Test Results

  • Appearance (colour):
    Red to very dark red
  • Appearance (Form):
    Liquid
  • Density:
    0.987 - 0.993
  • Wavelength (1)(UV):
    542 - 552 nm
  • Extinction (1)(UV):
    1%/1 cm
  • Solvent:
    Ethanol/H₂0 1/1

Specification Test Results

Related Information

Frequently Bought Together