L(-)-Menthol, Sigma-Aldrich®
: Merck
:2216-51-5
:
:C₁₀H₂₀O
:MFCD00062979
:156.27 g/mol
:13,05861
:212 °C (1013 hPa)
:41…44 °C
:0.89 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
:96 °C
:218-690-9
:GHS07
:Ambient
Frequently Bought Together