ContactUsLinkComponent
VWR® Rota® pH Indicator Papers
VWR® Rota® pH Indicator Papers
Catalog # VWRC109526.0005
Supplier:  VWR Chemicals
undefined
VWR® Rota® pH Indicator Papers
Catalog # VWRC109526.0005
Supplier:  VWR Chemicals
Specifications
  • pH gradation:
    1
  • pH range:
    1-10,0
  • Description:
    Universal Indicator, Rota
  • Pk:
    5
Specifications
About this item
These rolls (4,8 m × 10 mm) have only 1 colour zone and are useful where high precision of measurement is not necessary.