ContactUsLinkComponent
Microplates, standard height, 384 ShallowWell, Nunc™
Microplates, standard height, 384 ShallowWell, Nunc™
Catalog # 734-2877
   Low Manufacturing Impact
undefined
Microplates, standard height, 384 ShallowWell, Nunc™
Catalog # 734-2877
   Low Manufacturing Impact
Specifications
  • No. of wells:
    384
  • Recommended working volume:
    2 - 20 μl
  • Sterile:
    -
  • Colour:
    Black
  • Description:
    384 ShallowWell plate, non treated surface, without lid
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    NAR Certificate of Retirement
  • Pk:
    100
  • Well volume:
    25 μl
Specifications
About this item
Nunc™ 384 ShallowWell standard height, black and white plates save costs in samples and reagents by reducing the total well volume to 25 μl. The standard height of the plate allows the use of automation without making adjustments to robotics, readers or liquid handlers.