Water baths, WB series
Supplier: P D UMWELTSCHUTZ UND PRUEFGERA
Certificates
About this item
This range of high grade steel water baths has a variety of accessories to provide versatile combinations and enable various different applications to be performed. The temperature can be displayed in Fahrenheit or Celsius, and it is possible to calibrate using the actual value. Baths have a screw cap outlet to enable the water to be emptied, a drain tap can be ordered separately if required. Baths can be used with test tube racks, clamps for Erlenmeyer flasks.
- Easy operation via digital LED display
- Microprocessor control with adjustable over and under temperature limits, audible and visual warning signals
- Low liquid level protection to avoid any potential damage
- Infinitely adjustable shaking frequency from 20 to 200 rpm, and shaking amplitude of 16 mm when using optional
Certifications: Conform to DIN 12876-1 Class 1.
Ordering information: Supplied without accessories, accessories must be ordered separately.
Frequently Bought Together