ContactUsLinkComponent
Heating bath, stirred, HBR 4 control
Heating bath, stirred, HBR 4 control
Supplier:  IKA
Certificates
undefined
Heating bath, stirred, HBR 4 control
Supplier:  IKA
Specifications
  • Catalog No:
  • 462-0592
  • IKAA10001849
  • Capacity:
  • 4 L
  • 4 L
  • Temperature range:
  • RT*...200 °C
  • RT*...200 °C
  • Heating capacity:
  • 1 kW
  • 1 kW
  • Weight:
  • 4,4 kg
  • 4,4 kg