ContactUsLinkComponent
Thermosel systems
Thermosel systems
Catalog # 616-3193
Supplier:  AMETEK
undefined
Thermosel systems
Catalog # 616-3193
Supplier:  AMETEK
Specifications
  • Description:
    Thermosel system
  • Seq:
    02
  • Pk:
    1
Specifications
About this item
The Thermosel system provides a stable environment for viscosity measurements and rheological investigations up to 300 °C.
  • Temperature ramping between set points with Rheocalc software
  • Alignment bracket ensure concentricity of spindle and sample chamber
  • Disposable chambers available for difficult to clean samples
Suitable for use with hot melts such as asphalt, wax and polymers.
Delivery: Supplied as an SC4 spindle, alignment bracket, thermo container, sample chamber, five disposable sample chambers, extracting tools and RTD probe.