Automatic vacuum system, AA 02/AA 04
Supplier: HAEP LABOR
Certificates
About this item
Automatic safety vacuum system for extraction of surplus liquid.
- Suction power of 30 L air/min, absolute vacuum: 100 mbar
- A protection filter between the bottle and the vacuum pump protects the pump and its components
- The bottles are made from thick polypropylene
Consists of the following items:
Membrane pump with vacuum regulator and bottle holder in a compact unit
3 m suction hose ending in a pipette with a suction control button
Delivery: Comes with a powerful membrane pump with stainless steel housing mounted on a bottle safety holder.
Frequently Bought Together