Order Entry
Austria
ContactUsLinkComponent
Vacuum Systems LABOPORT® SC 820 and SC 840
Vacuum Systems LABOPORT® SC 820 and SC 840
Supplier:  KNF
Certificates
undefined
Vacuum Systems LABOPORT® SC 820 and SC 840
Supplier:  KNF

Specifications

  • Catalog No:
  • 181-3433
  • 181-3432
  • Model:
  • SCC 840
  • SCC 820
  • Vacuum pump:
  • N 840.3 FT.18
  • N 820.3 FT.18
  • Flow rate:
  • 34 l/min
  • 20 l/min
  • Working overpressure (bar):
  • 1,0
  • 1,0
  • Vacuum:
  • 8 mbar
  • 8 mbar
  • W×D×H:
  • 289×417×506 mm
  • 289×397×506 mm
  • Weight:
  • 20,9 kg
  • 17,6 kg

Specifications

Frequently Bought Together