Orders LinkContactUsLinkComponent
Replacement tubes
Replacement tubes
Supplier:   Vilber Lourmat
Replacement tubes
Supplier:  Vilber Lourmat
Certificates
Replacement tubes
23 Options Available Below
Product Details & Documents
Replacement tubes for transilluminators and UV lamps.
  • High UV irradiation and constant spectra
  • Longer shelf-life for thousands of irradiation hours
  • Unique electrode allowing for the highest reliability
Large output and wavelength ranges available.
Documents
Product Family Options
Boxes of 20 tubes
Product Information
  • Wavelength
    Output
    Length
    Availability
    Price
  • VILB907500611
    UV tubes, T-6.C
    UV tubes, T-6.C
    254 nm
    6 W
    211 mm
    Log in for availability
    Log in for availability
    - 347,00 €
    347,00 €
       347,00 €
  • VILB907500631
    UV tubes, T-6.L
    UV tubes, T-6.L
    365 nm
    6 W
    211 mm
    Log in for availability
    Log in for availability
    - 281,00 €
    281,00 €
       281,00 €
  • VILB907500811
    UV tubes, T-8.C
    UV tubes, T-8.C
    254 nm
    8 W
    288 mm
    Log in for availability
    Log in for availability
    - 361,00 €
    361,00 €
       361,00 €
  • VILB907500821
    UV tubes, T-8.M
    UV tubes, T-8.M
    312 nm
    8 W
    288 mm
    Log in for availability
    Log in for availability
    - 896,00 €
    896,00 €
       896,00 €
  • VILB907500831
    UV tubes, T-8.L
    UV tubes, T-8.L
    365 nm
    8 W
    288 mm
    Log in for availability
    Log in for availability
    - 269,00 €
    269,00 €
       269,00 €
  • VILB907501511
    UV tubes, T-15.C
    UV tubes, T-15.C
    254 nm
    15 W
    435 mm
    Log in for availability
    Log in for availability
    - 487,00 €
    487,00 €
       487,00 €
  • VILB907501521
    UV tubes, T-15.M
    UV tubes, T-15.M
    312 nm
    15 W
    435 mm
    Log in for availability
    Log in for availability
    - 1 070,00 €
    1 070,00 €
       1 070,00 €
  • VILB907501531
    UV tubes, T-15.L
    UV tubes, T-15.L
    365 nm
    15 W
    435 mm
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 306,00 €
    306,00 €
    1 * 20 ST
       306,00 €
Boxes of six tubes and starters
Product Information
  • Wavelength
    Output
    Length
    Availability
    Price
  • VILB907510811
    UV tubes T-8.C and starters
    UV tubes T-8.C and starters
    254 nm
    8 W
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 203,00 €
    203,00 €
    1 * 6 ST
       203,00 €
  • VILB907510821
    UV tubes T-8.M and starters
    UV tubes T-8.M and starters
    312 nm
    8 W
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 318,00 €
    318,00 €
    1 * 6 ST
       318,00 €
  • VILB907510831
    UV tubes T-8.L and starters
    UV tubes T-8.L and starters
    365 nm
    8 W
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 132,00 €
    132,00 €
    1 * 6 ST
       132,00 €
  • VILB907511511
    UV tubes T-15.C and starters
    UV tubes T-15.C and starters
    254 nm
    15 W
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 187,00 €
    187,00 €
    1 * 6 ST
       187,00 €
  • VILB907511521
    UV tubes T-15.M and starters
    UV tubes T-15.M and starters
    312 nm
    15 W
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 390,00 €
    390,00 €
    1 * 6 ST
       390,00 €
  • VILB907511531
    UV tubes T-15.L and starters
    UV tubes T-15.L and starters
    365 nm
    15 W
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 142,00 €
    142,00 €
    1 * 6 ST
       142,00 €
Single tubes
Product Information
  • Wavelength
    Output
    Length
    Availability
    Price
  • VILB907520411
    UV tubes, T-4.C
    UV tubes, T-4.C
    254 nm
    4 W
    135 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 34,70 €
    34,70 €
    1 * 1 ST
       34,70 €
  • VILB907520611
    UV tubes, T-6.C
    UV tubes, T-6.C
    254 nm
    6 W
    211 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 43,40 €
    43,40 €
    1 * 1 ST
       43,40 €
  • VILB907520631
    UV tubes, T-6.L
    UV tubes, T-6.L
    365 nm
    6 W
    211 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 40,50 €
    40,50 €
    1 * 1 ST
       40,50 €
  • VILB907520811
    UV tubes, T-8.C
    UV tubes, T-8.C
    254 nm
    8 W
    288 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 43,40 €
    43,40 €
    1 * 1 ST
       43,40 €
  • VILB907520821
    UV tubes, T-8.M
    UV tubes, T-8.M
    312 nm
    8 W
    288 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 86,70 €
    86,70 €
    1 * 1 ST
       86,70 €
  • VILB907520831
    UV tubes, T-8.L
    UV tubes, T-8.L
    365 nm
    8 W
    288 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 40,50 €
    40,50 €
    1 * 1 ST
       40,50 €
  • VILB907521511
    UV tubes, T-15.C
    UV tubes, T-15.C
    254 nm
    15 W
    435 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 54,90 €
    54,90 €
    1 * 1 ST
       54,90 €
  • VILB907521521
    UV tubes, T-15.M
    UV tubes, T-15.M
    312 nm
    15 W
    435 mm
    Log in for availability
    Log in for availability
    - 102,00 €
    102,00 €
       102,00 €
  • VILB907521531
    UV tubes, T-15.L
    UV tubes, T-15.L
    365 nm
    15 W
    435 mm
    Log in for availability
    Log in for availability
    - 43,40 €
    43,40 €
       43,40 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Starter, ST151 for 40 W and 20 W tube
    Starter, ST151 for 40 W and 20 W tube
    VILB907500011
    Log in for availability
    1 * 1 ST - 22,70 €
Product Information
  • Catalog #
    Availability
    Price
  • Starter, ST111 for 30 W and 40 W tube
    Starter, ST111 for 30 W and 40 W tube
    VILB907500021
    Log in for availability
    1 * 1 ST - 22,70 €