Accessories for Turbiquant®
Supplier: Merck
Certificates
About this item
In addition to spare cuvettes and light sources, the pour and flow cells have been developed to improve efficiency. The pour through assembly provides a single one-time cuvette alignment and simplified rinsing.
Continuous flow through measurement is possible with the Turbiquant 3000 T/3000 IR by using the flow assembly together with the periodically measured output via printer or PC.
Frequently Bought Together