Order Entry
Austria
ContactUsLinkComponent
VWR®, Digital Stopwatch
VWR®, Digital Stopwatch
Catalog # 609-0139
Supplier:  VWR Collection
undefined
VWR®, Digital Stopwatch
Catalog # 609-0139
Supplier:  VWR Collection

Specifications

  • Colour:
    Black
  • Description:
    Digital stopwatch, resolution 0,001
  • Seq:
    01
  • Accuracy:
    0,01 s
  • Pk:
    1
  • W×H:
    63×80 mm

Specifications

About this item

This electronic stopwatch includes a calendar and a clock.

  • 1/100 second chronograph
  • Alarm function
  • Memory function (stores one lap time)
Certifications: Tested and calibrated in a triple accredited/registered ISO/IEC 17025:2005, A2LA accredited laboratory (Cert #2448.01), against equipment whose calibration is traceable to NIST.
Delivery: Supplied with one CR2032 battery, certificate of calibration, individual serial number, and multi-language (English, French, German, Italian, Portuguese and Spanish) instructions.