High precision thermometers, TFN 520 and 530 series
Supplier: EBRO
Certificates
About this item
These thermometers can be used for measuring temperatures in air conditioning and ventilation applications, air and gases, liquids and for measuring surface temperatures, temperatures of tools for injection moulding processes and plastic materials.
- Maximum/minimum values displayed
- SMP, Lemo or Ex proof connections
- 'HOLD' function
- Automatic switch-off
- Response times (t99) of three seconds (620-1461)
External thermocouple types: K, J, T, E probes.
Standard version IP 52, SMP versions IP 40.
Weight: 90 g
Standard version IP 52, SMP versions IP 40.
Delivery: Supplied with three-point factory certificate of calibration, battery with charge indicator, without probe.
Frequently Bought Together