VWR® Traceable®, Wide Range Digital Thermometer
620-2004
- Range:−50…+700 °C
- Resolution:0,1 (–200…+640); 1,0 °C
- Weight:184 g
- W×D×H:70×32×178 mm
- Accuracy:±1,0 °C
- Pk:1
For quality control, routine monitoring, measuring temperatures in cuvettes, gas systems, reagents, chemical solutions, soil, water baths, ovens, incubators, petroleum products, foods and wastewater. Instant memory key recalls minimum and maximum readings over any time period. Stainless steel K-type thermocouple probe for any J-, K- or T-type probe.
- Computer output allows thermometer to be connected to a computer or data logger for capturing and storing readings
- Large, backlit LCD: Values can be read even in poor light
- Count-up timer provides a relative time measurement while taking temperatures and recording min./max. memories
NOTE: Values are valid for the included stainless steel probe. The thermometer can measure temperatures from –200 to +1370 °C with suitable probes that need to be ordered separately.
: Traceable to NIST plus conform to International Temperature Standard, ITS-90. Compliant with ISO/IEC 17025, ANSI/NCSL Z540-1 and 9001.
: Supplied with probe, 9 volt battery and Traceable® certificate of calibration.