Overhead stirrer shafts
: COWIE
PTFE, with encapsulated steel reinforced drive shaft. Chemically inert. The smooth surface reduces contamination and makes cleaning easier.
- Non scratch, prevents agitation vessels from being damaged
- Temperature resistant from −200 to +280 °C
: EX denotes exposed end, these stirrers have a stainless steel shaft core with an exposed end for rigid clamping.
Frequently Bought Together