Stirrers with stirrer bearing
Lieferant: LENZ LABORGLAS GMBH & CO.KG
Zertifikate
Über diesen Artikel
DURAN®, borosilicate glass, PTFE blade and securing screw. Gas tight closure for vacuum stirring apparatus up to 10⁻³ mbar at continuous operational speed of 800 min⁻¹.
- Ideal for use with 250 to 2000 ml round bottomed flasks
- Includes PP screw cap with GL 25 closure
Frequently Bought Together