VWR®, Stirrer Blades with Plain Ends
Supplier: VWR Collection
Certificates
About this item
PTFE. Blades fit PTFE stirring shaft; hole Ø 6,5 mm, 3,2 mm thick. Inert and non scratch. The smooth surface prevents contamination and makes cleaning easier.
- Excellent chemical resistance
- Temperature resistant from −200 to 280 °C
Frequently Bought Together