Order Entry
Austria
ContactUsLinkComponent
VWR®, Multi-Tube Vortexers
VWR®, Multi-Tube Vortexers
Supplier:  VWR Collection
Certificates
undefined
VWR®, Multi-Tube Vortexers
Supplier:  VWR Collection

Specifications

  • Catalog No:
  • 444-7061
  • 444-7063
  • Model:
  • Digital
  • Analogue
  • Orbit:
  • 3,6 mm
  • 3,6 mm
  • Speed:
  • 500 - 2500 min⁻¹
  • 1200 - 2400 min⁻¹
  • W×D×H:
  • 384×241×406 mm
  • 384×241×406 mm
  • Weight:
  • 18,6 kg
  • 18,6 kg
  • Timer:
  • 1 - 9999 min
  • 0 - 1 min
  • Motor:
  • 100 W
  • 100 W

Specifications

Frequently Bought Together