Vortex mixer, VORTEX Genius 3
About this item
Vortex mixer with three interchangeable attachments and various inserts for different applications. Microcentrifuge tubes, microtitre plates, even round bottomed or 250 ml Erlenmeyer flasks can be attached by means of the specially designed strap. Attachments click securely onto the unit in any position. The unit is stable at high speeds and sturdy with coated, zinc die-casting.
- Compact design
- Wide speed range, infinitely adjustable
- Touch or continuous mode
- Can be used at 5 to 40 °C (80% relative humidity) in an incubator or cold room
IP protection class according to DIN EN 60529: IP 21
* Depending on attachment and loading
Certifications: IP protection class according to DIN EN 60529: IP 21
Ordering information: Supplied with standard attachment for test tubes or small beakers, 412-0099.
Caution: * Depending on attachment and loading
** Use in combination with 412-0101
1 Options Available Below
Product Details & Documents
Vortex mixer with three interchangeable attachments and various inserts for different applications. Microcentrifuge tubes, microtitre plates, even round bottomed or 250 ml Erlenmeyer flasks can be attached by means of the specially designed strap. Attachments click securely onto the unit in any position. The unit is stable at high speeds and sturdy with coated, zinc die-casting.
- Compact design
- Wide speed range, infinitely adjustable
- Touch or continuous mode
- Can be used at 5 to 40 °C (80% relative humidity) in an incubator or cold room
IP protection class according to DIN EN 60529: IP 21
* Depending on attachment and loading
Certifications: IP protection class according to DIN EN 60529: IP 21
Ordering information: Supplied with standard attachment for test tubes or small beakers, 412-0099.
Caution: * Depending on attachment and loading
** Use in combination with 412-0101
Specifications for Product Family
Specifications
- Catalog No:
- 412-0098
- 412-0115
- 412-0116
- Model:
- VORTEX Genius 3
- VORTEX Genius 3
- VORTEX Genius 3
- Shaking motion:
- Orbital
- Orbital
- Orbital
- Orbit:
- 4 mm
- 4 mm
- 4 mm
- Speed range:
- 500 - 2500 min⁻¹*
- 500 - 2500 min⁻¹*
- 500 - 2500 min⁻¹*
- Motor input/output:
- 58/10 W
- 58/10 W
- 58/10 W
- W×D×H:
- 127×149×136 mm
- 127×149×136 mm
- 127×149×136 mm
- Weight:
- 4,5 kg
- 4,5 kg
- 4,5 kg
Specifications
Product Family Options
Product Information
- AvailabilityPrice
- 412-0098VORTEX Genius 3, EU-plugSpecifications:
More
Specifications
Cat. No.412-0098ModelVORTEX Genius 3Shaking motionOrbitalOrbit4 mmSpeed range500 - 2500 min⁻¹*Motor input/output58/10 WW×D×H127×149×136 mmWeight4,5 kg - 412-0115VORTEX Genius 3, UK-plugVORTEX Genius 3, UK-plugDiscontinued-Specifications:
More
Specifications
Cat. No.412-0115ModelVORTEX Genius 3Shaking motionOrbitalOrbit4 mmSpeed range500 - 2500 min⁻¹*Motor input/output58/10 WW×D×H127×149×136 mmWeight4,5 kg - 412-0116VORTEX Genius 3, CH-plugVORTEX Genius 3, CH-plugDiscontinued-Specifications:
More
Specifications
Cat. No.412-0116ModelVORTEX Genius 3Shaking motionOrbitalOrbit4 mmSpeed range500 - 2500 min⁻¹*Motor input/output58/10 WW×D×H127×149×136 mmWeight4,5 kg
Accessories
Product Information
- Catalog #ForAvailabilityPrice
- Flask attachment for Erlenmeyer or round bottom flasks, 100 up to 250 ml**Flask attachment for Erlenmeyer or round bottom flasks, 100 up to 250 ml**412-0107Continuous modeLog in for availability1 * 1 ST - 43,50 €
Product Information
- Catalog #ForAvailabilityPrice
- Test tube attachment, for 8 reagent tubes, 16 mm**Test tube attachment, for 8 reagent tubes, 16 mm**412-0105Continuous modeLog in for availability1 * 1 ST - 43,50 €
Product Information
- Catalog #ForAvailabilityPrice
- Test tube attachment, for 18 reagent tubes, 10 mm**Test tube attachment, for 18 reagent tubes, 10 mm**412-0103Continuous modeLog in for availability1 * 1 ST - 43,50 €
Product Information
- Catalog #ForAvailabilityPrice
- Test tube attachment, for 12 reagent tubes, 12 mm**Test tube attachment, for 12 reagent tubes, 12 mm**412-0104Continuous modeLog in for availability1 * 1 ST - 43,50 €
Product Information
- Catalog #ForAvailabilityPrice
- Test tube attachment, for 54 microcentrifuge tubes, 1,5/2,0 ml**Test tube attachment, for 54 microcentrifuge tubes, 1,5/2,0 ml**412-0102Continuous modeLog in for availability1 * 1 ST - 81,50 €
Product Information
- Catalog #ForAvailabilityPrice
- Test tube attachment, for 8 reagent tubes, 20 mm**Test tube attachment, for 8 reagent tubes, 20 mm**412-0106Continuous modeLog in for availability1 * 1 ST - 43,50 €
Product Information
- Catalog #ForAvailabilityPrice
- Microtitre plate attachment for 1 standard microtitre plate**Microtitre plate attachment for 1 standard microtitre plate**441-0036Continuous modeLog in for availability1 * 1 ST - 43,50 €
Product Information
- Catalog #ForAvailabilityPrice
- Attachment for single-handed operation, 88 mm round with rubber insertAttachment for single-handed operation, 88 mm round with rubber insert412-0100Continuous or touch modeLog in for availability1 * 1 ST - 31,50 €
Product Information
- Catalog #ForAvailabilityPrice
- Universal attachment, 150 mm with rubber insertUniversal attachment, 150 mm with rubber insert412-0101Continuous modeLog in for availability1 * 1 ST - 35,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Standard attachment
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



