Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Multifunction 3 dimensional rotator, PS-M3D

Multifunction 3 dimensional rotators, PS-M3D
Catalog # 444-0353
Supplier:   GRANT INSTRUMENTS
Multifunction 3 dimensional rotator, PS-M3D
Catalog # 444-0353
Supplier:  GRANT INSTRUMENTS
Multifunction 3 dimensional rotator, PS-M3D
Multifunction 3 dimensional rotator, PS-M3D

Multifunction 3 dimensional rotators, PS-M3D

Price
810,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

The PS-M3D is a fixed angle, variable speed, multifunction 3D rotator providing variable mixing options in a compact unit. Applications include immunoprecipitation, tissue culture treatments, gel staining, destaining, antibody staining, washes, hybridisations and blots. The stepper motor produces regulated and reproducible rotating throughout the speed range. The PS-M3D provides all that is needed for mixing different sized particles.

  • Multifunction: Rotation, reciprocation and vibration
  • All actions can be set for continuous or timed operation
  • Individual phases can be linked together for flexibility
  • Can be used at 4 to 40 °C (80% relative humidity, non-condensing), in an incubator or cold room

3D orbital (long wave) required for large particles.

Reciprocal (long and medium wave) ideal for large to medium particles.

Vibration (short wave) suits small particles.

Combining different phases in a customised sequence gives the following possible combinations: 3D orbital; 3D reciprocal; 3D vibration; 3D orbital and reciprocal; 3D orbital and vibration; 3D orbital and reciprocal and vibration; 3D reciprocal and vibration. An individual program may be configured for any different combination of components to ensure optimum mixing conditions.

Delivery information: Supplied with non-slip mat, optional dimpled mat is available separately.

Caution: Working area: 200×200 mm

Specifications
  • Description:
    Multifunction 3 dimensional rotators, PS-M3D
  • Speed:
    1 - 100 min⁻¹
  • Turning angle - vibration mode:
    1 - 5°
  • Input power:
    4,6 W (External power supply: 12 V, 500 mA)
  • No. of cycles:
    1 - 125
  • Platform W×D:
    235×235 mm*
  • Weight:
    2 kg
  • Shaking motion:
    3D Orbital, reciprocal, vibration
  • Cat. No.:
    444-0353
  • Angle:
  • Timer:
    1 - 250 s (360°/reciprocal mode)
    1 - 5 s (360°/vibration mode)
  • Model:
    PS-M3D
  • W×D×H:
    225×170×125 mm
  • Max. load:
    1 kg
  • Turning angle - reciprocal mode:
    1 - 360° (in steps of 30°)
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Dimpled mat
    Dimpled mat
    444-0338
    Log in for availability
    1 * 1 ST - 47,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.