Order Entry
Austria
ContactUsLinkComponent
Platform rocker, PMR-30
Platform rocker, PMR-30
Catalog # 444-0341
Supplier:  GRANT INSTRUMENTS
undefined
Platform rocker, PMR-30
Catalog # 444-0341
Supplier:  GRANT INSTRUMENTS

Specifications

  • Angle:
  • Shaking motion:
    Rocking, side to side (2D)
  • Description:
    PMR-30 platform rocker
  • Seq:
    01
  • Model:
    PMR-30
  • Max. load:
    1 kg
  • Input power:
    3,8 W (External power supply: 12 V; 0,32 A)
  • Platform W×D:
    210×210 mm
  • Speed rocking:
    5 - 30 cycles/min
  • Weight:
    2 kg
  • W×D×H:
    220×205×120 mm
  • Pk:
    1

Specifications

About this item

The compact and highly functional PMR-30 variable speed, fixed angle platform rocker is ideally suited for personal use, it provides a smooth side to side rocking motion for gentle sample agitation in tubes, culture flasks, dishes and boxes. Applications include gel staining, destaining, antibody staining, washes, hybridisations, and blots. The reliable motor with direct drive mechanism produces regulated and reproducible rocking throughout the speed range.

  • Continuous or timer operation; integral digital timer 1 min to 24 h, with automatic switch-off, time remaining is displayed
  • Can be used at 4 to 40 °C (80% relative humidity, non-condensing), in an incubator or cold room
Delivery: Supplied with non-slip mat, optional dimpled mat is available separately.