Orders LinkContactUsLinkComponent
Core samplers, QuickPicker
Core samplers, QuickPicker
Supplier:   BURKLE
Core samplers, QuickPicker
Supplier:  BURKLE
Zertifikate
Core samplers, QuickPicker
2 Optionen unten verfügbar
Product Details & Documents
Made of stainless steel or PP, transparent, sample bottles made of PE. The QuickPicker is appropriate when bulk goods are withdrawn directly from sacks, bags or open containers. The sample is filled directly into sample bottles. Sampling depth 300 mm, Ø 25 mm, length 500 mm, 75 ml volume.
  • Removable tip
  • Easy to handle and clean
  • No edges or crevices, no residual sample material
Delivery information: QuickPicker is delivered complete with two sample bottles and a cleaning brush.
Product Family Options
Produktinformationen
  • Length
    Immersion depth
    Ø
    Volume
    Availability
    Price
  • 312-1175
    Sampler, stainless steel V4A (1.4404)/AISI 316 L
    Sampler, stainless steel V4A (1.4404)/AISI 316 L
    500 mm
    300 mm
    25 mm
    75 ml
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 1 ST
    1 * 1 ST - 258,00 €
    258,00 €
    1 * 1 ST
       258,00 €
  • 312-0085
    Sampler, PP, transparent
    Sampler, PP, transparent
    500 mm
    300 mm
    25 mm
    75 ml
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 1 ST
    1 * 1 ST - 163,00 €
    163,00 €
    1 * 1 ST
       163,00 €
Accessories
Produktinformationen
  • Catalog #
    Capacity
    Thread
    Material
    Availability
    Price
  • 300-0118
    250 ml
    40 mm
    LDPE
    Einloggen für Verfügbarkeit
    1 * 1 ST - 2,40 €
Produktinformationen
  • Catalog #
    Availability
    Price
  • Transport case
    Transport case
    312-0071
    Einloggen für Verfügbarkeit
    1 * 1 ST - 36,30 €
Produktinformationen
  • Catalog #
    Capacity
    Thread
    Material
    Availability
    Price
  • 331-2612
    250 ml
    40 mm
    PP
    Einloggen für Verfügbarkeit
    1 * 1 ST - 2,70 €