Orders LinkContactUsLinkComponent
Zone samplers
 :   BURKLE
Zone samplers
 :  BURKLE
Zone samplers
Zone samplers
For sampling bulk goods, cross-sectional samples can be taken from different sample depths. Suitable for use with a range of sample substrates from fine powder to coarse grain. Multi sampler has three or five separate chambers for taking samples at multiple depths in one action. Uno sampler has one chamber for taking a sample at one specified depth. All layer sampler has no chambers, three or five inlets and this allows one sample to be taken at multiple depths. Jumbo sampler has no chambers and three, five or seven inlets; with a 50 mm tube diameter this is ideal for larger grain sizes and quantities.
  • Easy to clean, removable tip
  • Colour coding (Multi and Uno samplers)
  • Ultra-pure materials: Stainless steel V4A (1.4404)/AISI 316 L, PTFE or aluminium
  • 3 lengths from 550 mm to 1500 mm, Jumbo up to 2500 mm
  • Samplers have smallest possible gap between inner and outer tubes
Product Family Options
All layer samplers
  • Description
    Length
    Immersion depth
    No. of chambers
    Ø
    Volume
  • All layer sampler, aluminium
    550 mm
    430 mm
    3
    25 mm
    70 ml
    1 * 1 ST
    1 * 1 ST - 250,00 €
    250,00 €
    1 * 1 ST
       250,00 €
  • All layer sampler, aluminium
    1500 mm
    1355 mm
    5
    25 mm
    260 ml
    1 * 1 ST
    1 * 1 ST - 353,00 €
    353,00 €
    1 * 1 ST
       353,00 €
  • All layer sampler, aluminium
    850 mm
    710 mm
    3
    25 mm
    130 ml
    1 * 1 ST
    1 * 1 ST - 278,00 €
    278,00 €
    1 * 1 ST
       278,00 €
  • All layer sampler, V4A
    550 mm
    430 mm
    3
    25 mm
    70 ml
    1 * 1 ST
    1 * 1 ST - 669,00 €
    669,00 €
    1 * 1 ST
       669,00 €
  • All layer sampler, V4A
    850 mm
    710 mm
    3
    25 mm
    130 ml
    1 * 1 ST
    1 * 1 ST - 771,00 €
    771,00 €
    1 * 1 ST
       771,00 €
  • All layer sampler, V4A
    1500 mm
    1355 mm
    5
    25 mm
    260 ml
    1 * 1 ST
    1 * 1 ST - 928,00 €
    928,00 €
    1 * 1 ST
       928,00 €
Multi samplers
  • Description
    Length
    Immersion depth
    No. of chambers
    Ø
    Volume
  • Multi sampler, aluminium
    1500 mm
    1355 mm
    5
    25 mm
    17 ml
    1 * 1 ST
    1 * 1 ST - 408,00 €
    408,00 €
    1 * 1 ST
       408,00 €
  • Multi sampler, aluminium
    550 mm
    430 mm
    3
    25 mm
    14 ml
    1 * 1 ST
    1 * 1 ST - 284,00 €
    284,00 €
    1 * 1 ST
       284,00 €
  • Multi sampler, aluminium
    850 mm
    710 mm
    3
    25 mm
    17 ml
    1 * 1 ST
    1 * 1 ST - 316,00 €
    316,00 €
    1 * 1 ST
       316,00 €
  • Multi sampler, V4A/PTFE
    550 mm
    430 mm
    3
    25 mm
    14 ml
    1 * 1 ST
    1 * 1 ST - 487,00 €
    487,00 €
    1 * 1 ST
       487,00 €
  • Multi sampler, V4A/PTFE
    850 mm
    710 mm
    3
    25 mm
    17 ml
    1 * 1 ST
    1 * 1 ST - 546,00 €
    546,00 €
    1 * 1 ST
       546,00 €
  • Multi sampler, V4A
    1500 mm
    1355 mm
    5
    25 mm
    17 ml
    1 * 1 ST
    1 * 1 ST - 989,00 €
    989,00 €
    1 * 1 ST
       989,00 €
  • Multi sampler, V4A/PTFE
    1500 mm
    1355 mm
    5
    25 mm
    17 ml
    1 * 1 ST
    1 * 1 ST - 798,00 €
    798,00 €
    1 * 1 ST
       798,00 €
  • Multi sampler, V4A
    550 mm
    430 mm
    3
    25 mm
    14 ml
    1 * 1 ST
    1 * 1 ST - 730,00 €
    730,00 €
    1 * 1 ST
       730,00 €
  • Multi sampler, V4A
    850 mm
    710 mm
    3
    25 mm
    17 ml
    1 * 1 ST
    1 * 1 ST - 782,00 €
    782,00 €
    1 * 1 ST
       782,00 €
Uno samplers
  • Description
    Length
    Immersion depth
    No. of chambers
    Ø
    Volume
  • Uno sampler, V4A/PTFE
    550 mm
    430 mm
    1
    25 mm
    17 ml
    1 * 1 ST
    1 * 1 ST - 496,00 €
    496,00 €
    1 * 1 ST
       496,00 €
  • Uno sampler, V4A/PTFE
    850 mm
    710 mm
    1
    25 mm
    17 ml
    1 * 1 ST
    1 * 1 ST - 556,00 €
    556,00 €
    1 * 1 ST
       556,00 €
  • Uno sampler, V4A/PTFE
    1500 mm
    1355 mm
    1
    25 mm
    17 ml
    1 * 1 ST
    1 * 1 ST - 814,00 €
    814,00 €
    1 * 1 ST
       814,00 €
Jumbo samplers
  • Description
    Length
    Immersion depth
    No. of chambers
    Ø
    Volume
  • Jumbo sampler, all layer, open inner tube, aluminium
    850 mm
    710 mm
    3
    50 mm
    880 ml
    1 * 1 ST
    1 * 1 ST - 542,00 €
    542,00 €
    1 * 1 ST
       542,00 €
  • Jumbo sampler, all layer, open inner tube, aluminium
    1500 mm
    1355 mm
    5
    50 mm
    1700 ml
    1 * 1 ST
    1 * 1 ST - 768,00 €
    768,00 €
    1 * 1 ST
       768,00 €
  • Jumbo sampler, all layer, open inner tube, aluminium
    2500 mm
    2355 mm
    7
    50 mm
    2900 ml
    1 * 1 ST
    1 * 1 ST - 1 030,00 €
    1 030,00 €
    1 * 1 ST
       1 030,00 €
  • Jumbo sampler, multi, closed chambers, aluminium/PTFE
    850 mm
    710 mm
    3
    50 mm
    390 ml
    1 * 1 ST
    1 * 1 ST - 838,00 €
    838,00 €
    1 * 1 ST
       838,00 €
  • Jumbo sampler, multi, closed chambers, aluminium/PTFE
    1500 mm
    1355 mm
    5
    50 mm
    650 ml
    1 * 1 ST
    1 * 1 ST - 985,00 €
    985,00 €
    1 * 1 ST
       985,00 €
  • Jumbo sampler, multi, closed chambers, aluminium/PTFE
    2500 mm
    2355 mm
    7
    50 mm
    910 ml
    1 * 1 ST
    1 * 1 ST - 1 400,00 €
    1 400,00 €
    1 * 1 ST
       1 400,00 €
Accessories
  •  
    Description
    Ø
  • 300-0114
    Sealing sleeves, PP, sterilisable, Ø = 25 mm, 150 mm
    25 mm
    1 * 3 ST - 32,60 €
  •  
    Description
  • 300-0110
    Cleaning brush, PVC/stainless steel, Ø: 50 mm, 1000 mm
    1 * 1 ST - 59,00 €
  •  
    Description
  • 300-0112
    Quali-Paper for emptying collected samples, 890×230 mm
    1 * 50 ST - 42,80 €
  •  
    Description
  • 331-2172
    Cleaning brush, PVC/stainless steel, Ø: 25 mm, 800 mm
    1 * 1 ST - 53,70 €
  •  
    Description
  • 331-2173
    Cleaning brush, PVC/stainless steel, Ø: 25 mm, 600 mm
    1 * 1 ST - 53,70 €
  •  
    Description
    Ø
  • 300-0113
    Sealing sleeves, PP, sterilisable, Ø = 50 mm, 150 mm
    50 mm
    1 * 1 ST - 27,90 €
  •  
    Description
  • 300-0109
    Cleaning brush, PVC/stainless steel, Ø 25 mm, 400 mm
    1 * 1 ST - 53,70 €
  •  
    Description
  • 331-2170
    Cleaning brush, PVC/stainless steel, Ø: 25 mm, 1000 mm
    1 * 1 ST - 53,70 €
  •  
    Description
  • 312-1141
    Transport case
    1 * 1 ST - 135,00 €
  •  
    Description
  • 300-0059
    Stainless steel V2A emptying funnel
    1 * 1 ST - 73,90 €
  •  
    Description
  • 300-0111
    Cleaning brush, PVC/stainless steel, Ø: 50 mm, 3000 mm
    1 * 1 ST - 121,00 €
  •  
    Description
  • 300-0058
    Stainless steel V2A emptying device with three funnels, 1000 mm
    1 * 1 ST - 475,00 €