Orders LinkContactUsLinkComponent
VWR®, Sample Dippers
VWR®, Sample Dippers
Supplier:   VWR Collection
VWR®, Sample Dippers
Supplier:  VWR Collection
Certificates
VWR®, Sample Dippers
VWR®, Sample Dippers
5 Options Available Below
Product Details & Documents
These sample dippers feature a basic container with a screw-in PTFE encapsulated steel shaft.
  • Excellent chemical resistance, temperature resistant
  • The basic unit consists of a container with a single 600 mm shaft
  • Additional shafts (331-0005) can be screwed onto the handle to increase length
Suitable for use at high and low temperatures (–200 to +280 °C).
Product Family Options
Product Information
  • Material
    Length
    Ø
    Capacity
    Availability
    Price
  • 331-0006
    Sample dipper
    Sample dipper
    PTFE
    600 mm
    54 mm
    100 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 159,00 €
    159,00 €
    1 * 1 ST
       159,00 €
  • 331-0007
    Sample dipper
    Sample dipper
    PTFE
    600 mm
    66 mm
    250 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 199,00 €
    199,00 €
    1 * 1 ST
       199,00 €
  • 331-0008
    Sample dipper
    Sample dipper
    PTFE
    600 mm
    80 mm
    500 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 236,00 €
    236,00 €
    1 * 1 ST
       236,00 €
  • 331-0009
    Sample dipper
    Sample dipper
    PTFE
    600 mm
    100 mm
    1000 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 265,00 €
    265,00 €
    1 * 1 ST
       265,00 €
Replacement shaft
Product Information
  • Material
    Length
    Ø
    Capacity
    Availability
    Price
  • 331-0005
    Replacement/extension shaft
    Replacement/extension shaft
    PTFE
    600 mm
    -
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 106,00 €
    106,00 €
    1 * 1 ST
       106,00 €