Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Dipping vessel

Supplier:   BURKLE
Dipping vessel
Supplier:  BURKLE
Certificates
Dipping vessel

2 Options Available Below

Product Details & Documents

Dipping vessel for taking all-layer samples from tanks, tank wagons, water courses and water purification plants. Made of non spark-producing brass or stainless steel AISI 304 (1,4301) for Ex area.

  • Easy-Flow, flow optimised flow valve
  • High weight (2,1 kg) for faster lowering
  • Height with handles 427 mm
  • Tested for use in the Ex protection zone 0
  • Explosion Categories IIA, IIB and IIC

 : Please order the appropriate lowering cable at the same time.

Product Family Options

Product Information

  • Capacity
    Availability
    Price
  • 300-1033
    Dipping vessel, brass
    Dipping vessel, brass
    1000 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 644,00 €
    644,00 €
    1 * 1 ST
       644,00 €

  • 300-1034
    Dipping vessel, stainless steel AISI 304
    Dipping vessel, stainless steel AISI 304
    1000 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 270,00 €
    1 270,00 €
    1 * 1 ST
       1 270,00 €