Orders LinkContactUsLinkComponent
Sampler, UniSampler 'Ex'
UniSampler "Ex"
Catalog # 300-1044
Supplier:   BURKLE
Sampler, UniSampler 'Ex'
Catalog # 300-1044
Supplier:  BURKLE
Sampler, UniSampler 'Ex'
Sampler, UniSampler 'Ex'
UniSampler "Ex"
Price
434,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Glass bottle and hose are connected in such a way that they conduct electricity and are grounded via a grounding cable. Operation: Ground the device, immerse the suction hose to the required depth, attach the adapter to the sampling bottle, generate a vacuum with the manual pump and collect the sample, remove the filled sampling bottle, close it and label it.
  • Specially developed for sampling flammable liquids of hazardous materials (e.g. gasoline)
  • PTB approval D- 333-009 618/00
Specially developped for sampling flammable liquids of hazardous materials such as gasoline.
 : PTB approval D- 333-009 618/00.
 : Supplied complete with a four-meter hose, glass bottle, vacuum pump and grounding cable in transport case.
Specifications
  • Description:
    UniSampler "Ex"