Orders LinkContactUsLinkComponent
Sampler, Vampire
Vampire Sampler, portable sampler for liquids
  300-1024
 :   BURKLE
Sampler, Vampire
  300-1024
 :  BURKLE
Sampler, Vampire
Sampler, Vampire
Vampire Sampler, portable sampler for liquids
1 160,00 €
1 * 1 ST
The Vampire sampler can be used to take samples from liquids easily, even from deeper depths.
  • Battery-powered hose pump for sampling liquids from depth
  • Easy cleaning and exchangeable hoses prevent impurities
  • Suction depth up to 5 meters
  • Portable sampler is easy to handle and carry
  • Pump attachment is easy to attach to the drive unit
  • Sample bottle is fastened directly onto the sampling instrument by a bottle basket
The portable pump head consists of a mains-independent drive unit, a pump head, and a delivery hose.
The medium being sampled is directed via the closed hose system directly into the sample container. The sample only comes into contact with the hoses and the sample bottle. This excludes the risk of impurities and contamination because the hoses are easy to clean and can also be replaced easily if necessary.
The self-priming peristaltic pump is also suitable for substances that are dirty or contain particles with a maximum viscosity of 500 mPas. The gentle pumping does not change the sample. Thanks to a suction depth of 5 meters, the battery-powered Vampire Sampler easily pumps samples, even from sampling points at lower depths, for example from barrels, tanks, wells, water traps, or wastewater and sewage treatment plants.
The hose pump is perfect for sampling liquid substances in industrial applications such as beverage production or monitoring water quality for environmental protection.
 : Consisting of a drive unit, pump head, pump hose (30 cm, Marprene), suction hose (5 m, PE), hose connector (PP), sample bottle (250 ml, LDPE), bottle basket, two rechargeable batteries and battery charger.
  • Description:
    Vampire Sampler, portable sampler for liquids
Accessories
  •  
    Description
  • 300-1045
    Suction hose, PE, Ø: 6 mm, thickness = 1 mm
    1 * 100 m - 87,40 €
  •  
    Capacity
    Thread
    Material
  • 300-0174
    500 ml
    50 mm
    LDPE
    1 * 1 ST - 3,90 €
  •  
    Description
  • 228-1191
    Suction hose, PE, Ø: 6 mm, thickness = 1 mm
    1 * 10 m - 23,00 €
  •  
    Description
  • 300-0176
    Suction hose, marprene, 1 m
    1 * 1 m - 60,30 €
  •  
    Description
  • BURK5302-2025
    Basket for 250 ml sample bottles, 68 mm
    1 * 1 ST - 54,30 €
  •  
    Description
  • BURK5302-2051
    Basket for 500 ml sample bottles, 84 mm
    1 * 1 ST - 67,60 €
  •  
    Description
  • BURK5302-2101
    Basket for 1000 ml sample bottles, 100 mm
    1 * 1 ST - 93,00 €