ContactUsLinkComponent
Pharmaceutical refrigerator/freezer combinations, PHCbi
Pharmaceutical refrigerator/freezer combinations, PHCbi
Supplier:  PHC Corporation
Certificates
undefined
Pharmaceutical refrigerator/freezer combinations, PHCbi
Supplier:  PHC Corporation
Specifications
  • Catalog No:
  • 472-0011
  • Model:
  • MPR-414F
  • Cat. No.:
  • 472-0011
  • Temperature:
  • +2...+14/-20...-30 °C
  • Display:
  • LED
  • Alarm:
  • auditive and visual
  • Access ports:
  • 2
  • Shelves/drawers:
  • 5 shelves (refrigerator), 1 shelf (freezer)
  • Door type:
  • 4 solid swing doors
  • Door lock:
  • Yes
  • Material:
  • stainless steel/coloured aluminium
  • Specifications:
  • with 4 castors and 2 adjustable feet
  • Weight:
  • 126 kg