ContactUsLinkComponent
Commercial refrigerators with forced air cooling
 :  Liebherr
undefined
Commercial refrigerators with forced air cooling
 :  Liebherr
  • Catalog No:
  • 471-1180
  • 471-1036
  • LIEBFKUV1660
  • 471-0471
  • Model:
  • FKUv 1663
  • FKv 503
  • FKUv 1660
  • FKUv 1610
  • Cat. No.:
  • 471-1180
  • 471-1036
  • LIEBFKUV1660
  • 471-0471
  • Temperature:
  • +1...+15 °C
  • +1...+12 °C
  • 1...15 °C
  • +1...+15 °C
  • Alarm:
  • Audible and visual door and temperature alarm
  • -
  • Audible and visual door and temperature alarm
  • Audible and visual door and temperature alarm
  • Shelves/drawers:
  • 3 shelves
  • 2 shelves
  • 3 shelves
  • 3 shelves
  • Door type:
  • 1 glass swing door
  • 1 glass swing door
  • 1 solid swing door
  • 1 solid swing door
  • W×D×H ext.:
  • 600×615×830 mm
  • 425×450×612 mm
  • 600×615×830 mm
  • 600×615×830 mm
  • Weight:
  • 42 kg
  • 31 kg
  • 35 kg
  • 35 kg
Frequently Bought Together