Orders LinkContactUsLinkComponent
Handheld refactometers
Handheld refactometers
Supplier:   Mettler - Toledo
Handheld refactometers
Supplier:  Mettler - Toledo
Certificates
Handheld refactometers
2 Options Available Below
Product Details & Documents
These refractometers, which use the total reflection method, are handheld instruments for measurements in the laboratory and production environments. The backlit LC display and the simple intuitive menu structure allow the user to easily change settings in seconds. The sample can either be placed on the measurement cell or the sensor can be immersed directly into the fluid.
  • Selectable automatic temperature compensation (ATC)
  • Handheld or benchtop instrument
  • Choice of use: Immerse sensor in liquid or add sample to cell
  • Storage of up to 1100 results and easy transfer with IR interface (adapter accessory 635-0117 required)
  • Refracto GS has sapphire prism and thus extended range and faster response
The results can be displayed in a wide variety of standard units and also units applicable to antifreeze and wine measurement as well as customised applications.
Delivery information: Supplied with carry case, HelloCD™ software, two AAA batteries, cleaning tissues, two LDPE pipettes, two PE vials and operating instructions.
Specifications for Product Family
Specifications
  • Catalog No:
  • 635-0445
  • 635-0446
  • Response time:
  • 3 - 180 seconds
  • 3 - 180 seconds
  • Interfaces:
  • Infrared for printer or PC
  • Infrared for printer or PC
  • Operating temperature:
  • 10...40 °C
  • 10...40 °C
  • Electrical:
  • Two AAA batteries
  • Two AAA batteries
  • Weight:
  • 250 g
  • 250 g
Specifications
Product Family Options
Product Information
  • Model
    Range
    Precision
    Resolution
    Availability
    Price
  • 30PX
    0 - 85 °Bx ; 1,32 - 1,50 nD
    0,1 °Bx ; ±0,0005 nD
    0,0001 nD
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3 140,00 €
    3 140,00 €
    1 * 1 ST
       3 140,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    635-0445
    Response time
    3 - 180 seconds
    Interfaces
    Infrared for printer or PC
    Operating temperature
    10...40 °C
    Electrical
    Two AAA batteries
    Weight
    250 g
  • 30GS
    0 - 85 °Bx ; 1,32 - 1,62 nD
    0,1 °Bx ; ±0,0005 nD
    0,0001 nD
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4 120,00 €
    4 120,00 €
    1 * 1 ST
       4 120,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    635-0446
    Response time
    3 - 180 seconds
    Interfaces
    Infrared for printer or PC
    Operating temperature
    10...40 °C
    Electrical
    Two AAA batteries
    Weight
    250 g
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Infrared adapter / RS232 DB9M
    Infrared adapter / RS232 DB9M
    635-0117
    Log in for availability
    1 * 1 ST - 700,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Cleaning tissues
    Cleaning tissues
    METT51325003
    Log in for availability
    1 * 10 ST - 39,50 €