Complete peristaltic pump sets
Supplier: Heidolph Instruments GmbH & Co.KG
Certificates
About this item
Complete sets including pump drive, pump head, and tubing. Ready to use.
- Rated flow 0,3 to 80 ml/min
- For tubing with 1,6 mm wall thickness
- The tubing set includes 1 m Tygon tubing (internal diameter 0,8 mm) and 1 m silicone tubing (internal diameter 0,8 mm)
Peristaltic pump drive PD 5201 (for technical description see item 224-1354) with SP Quick 1,6 pump head and tubing set.
Peristaltic pump drive PD 5206 (for technical description see item 224-1355) with SP Standard 2,5 pump head and tubing set.
Frequently Bought Together