Complete peristaltic pump sets
Complete sets consisting of tube drive, pump head, and tubing. Ready-to-use.
- Rated flow 0,3 to 80 ml/min
- For tubing with 1,6 mm wall thickness
- The tubing set includes 1 m Tygon tubing (internal diameter 0,8 mm) and 1 m silicone tubing (internal diameter 0,8 mm)
Peristaltic pump drive PD 5101 (for technical description see item 224-1352) with SP Quick 1,6 pump head and tubing set.
Frequently Bought Together