Hei-FLOW Silver 1, Hei-FLOW Silver 2 Peristaltic Pump Packages
Packages for Hei-FLOW Value 01 Hei-FLOW Value 06 peristaltic pumps.
- For standard applications and liquid transfer
Hei-FLOW Silver 1 Package includes Hei-FLOW Value 01 (model PD 5001), Pump head SP quick 1,6, and 1 m each Tygon® (standard) and Silicone tubing (inner Ø 3,1 mm).
Hei-FLOW Silver 2 Package includes Hei-FLOW Value 06 (model PD 5006), Pump head SP standard 2,5, and 1 m each Tygon® (standard) and Silicone tubing (inner Ø 6,4 mm).
: For more information, see peristaltic pump drive models PD 5001 and PD 5006.
Frequently Bought Together